Capric acid ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.02169.0100
8.02169.1000
8.02169.9010
8.02169.0100
8.02169.1000
8.02169.9010
EM8.02169.0100CS
1058.6
CAD
EM8.02169.0100
EM8.02169.1000
EM8.02169.9010
CA8.02169.0100
CA8.02169.1000
CA8.02169.9010
Capric acid ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Capric acid
Formula:
C₁₀H₂₀O₂ Boiling Pt: 268 °C (1013 hPa) Melting Pt: 29…32 °C Density: 0.89 g/cm3 (20 °C) Flash Pt: 147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004441 CAS Number: 334-48-5 EINECS: 206-376-4 Merck Index: 13,01764 |
Specification Test Results
Solubility | 0.15 g/l (20 °C) |
Melting Point | 29 - 32 °C |
Molar Mass | 172.26 g/mol |
Bulk Density | 690 kg/m3 |
Boiling Point | 268 - 270 °C |
Vapor Pressure | 0.13 hPa (79 °C) |
Flash Point | 147 °C |
Refractive Index | 1.428 (40 °C, 589 nm) |
Density | 0.89 g/cm3 (20 °C) |
pH | 4 (0.2 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...