Benzonitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Cyanobenzene, Phenylcyanide

M-Clarity™ quality level = MQ200

8.01800.0100 8.01800.0500 8.01800.1000 8.01800.0100 8.01800.0500 8.01800.1000
EM8.01800.0100CS 826.34 CAD
EM8.01800.0100 EM8.01800.0500 EM8.01800.1000 CA8.01800.0100 CA8.01800.0500 CA8.01800.1000
Benzonitrile for synthesis, Sigma-Aldrich®
Benzonitrile
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 190.7
Melting Pt: -13
Density: 1 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1 II
Merck Index: 13,01098

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Melting Point -13 °C
Molar Mass 103.12 g/mol
Boiling Point 190 °C (1013 hPa)
Vapor Pressure 0.7 hPa (20 °C)
Flash Point 70 °C
Refractive Index 1.5289 (20 °C, 589 nm)
Explosion Limit 1.4 - 7.2 %(V)
Density 1.00 g/cm3 (20 °C)
Ignition Point 550 °C (experimental)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR