1-Bromopropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Solvent 38, n-Propyl bromide

M-Clarity™ quality level = MQ200

8.01667.0005 8.01667.0250 8.01667.1000 8.01667.0005 8.01667.0250 8.01667.1000
EM8.01667.0005CS 2880.34 CAD
EM8.01667.0005 EM8.01667.0250 EM8.01667.1000 CA8.01667.0005 CA8.01667.0250 CA8.01667.1000
1-Bromopropane for synthesis, Sigma-Aldrich®
1-Bromopropane
Formula: BrH₂CCH₂CH₃
MW: 122.99 g/mol
Boiling Pt: 71
Melting Pt: –108 °C
Density: 1.354 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00000254
CAS Number: 106-94-5
EINECS: 203-445-0
UN: 2344
ADR: 3 II
Merck Index: 12,08030
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e928c183f1c573c1a2eba4ccb7f1147af103ed&vid=6b2b626154a7592a42b400fdf8ba1c1028a60850&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 2.5 g/l (20 °C)
Melting Point -108 °C
Molar Mass 122.99 g/mol
Boiling Point 71 °C (1013 hPa)
Vapor Pressure 146 hPa (20 °C)
Flash Point -10 °C
Refractive Index 1.4341 (20 °C, 589 nm)
Explosion Limit 3.4 - 9.1 %(V)
Density 1.35 g/cm3 (20 °C)
Ignition Point 490 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR