Benzil for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,2-Diphenyl-1,2-ethanedione

M-Clarity™ quality level = MQ200

8.01632.0005 8.01632.0250 8.01632.0005 8.01632.0250
EM8.01632.0005CS 51865.08 CAD
EM8.01632.0005 EM8.01632.0250 CA8.01632.0005 CA8.01632.0250
Benzil for synthesis, Sigma-Aldrich®
Benzil
Formula: (C₆H₅)C(O)C(O)(C₆H₅)
MW: 210.23 g/mol
Boiling Pt: 346
Melting Pt: 95
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003080
CAS Number: 134-81-6
EINECS: 205-157-0
Merck Index: 13,01078

Order Now

Specification Test Results

Solubility  - 0.5 g/l (20 °C)
Melting Point 95 °C
Molar Mass 210.23 g/mol
Boiling Point 346 - 348 °C
Vapor Pressure 1.3 hPa (128 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR