1-Bromohexane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1-Hexyl bromide

M-Clarity™ quality level = MQ200

8.01603.0100 8.01603.0500 8.01603.0100 8.01603.0500
EM8.01603.0100CS 1014.36 CAD
EM8.01603.0100 EM8.01603.0500 CA8.01603.0100 CA8.01603.0500
1-Bromohexane for synthesis, Sigma-Aldrich®
1-Bromohexane
Formula: BrH₂C(CH₂)₄CH₃
MW: 165.07 g/mol
Boiling Pt: 155
Melting Pt: –85 °C
Density: 1.167 g/cm³ (20 °C)
Flash Pt: 47
Storage Temperature: Ambient
MDL Number: MFCD00000271
CAS Number: 111-25-1
EINECS: 203-850-2
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Solubility  - 1 g/l (20 °C)
Melting Point -85 °C
Molar Mass 165.07 g/mol
Boiling Point 154 - 158 °C
Vapor Pressure 4 hPa (20 °C)
Flash Point 47 °C
Refractive Index 1.4478 (20 °C, 589 nm)
Density 1.18 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR