1-Bromo-2-methylpropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01549.0250 8.01549.1000 8.01549.0250 8.01549.1000
EM8.01549.0250CS 785.26 CAD
EM8.01549.0250 EM8.01549.1000 CA8.01549.0250 CA8.01549.1000
1-Bromo-2-methylpropane for synthesis, Sigma-Aldrich®
1-Bromo-2-methylpropane
Formula: C₄H₉Br
Boiling Pt: 91 °C (1013 hPa)
Melting Pt: –118 °C
Density: 1.265 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000217
CAS Number: 78-77-3
EINECS: 201-141-2
UN: 2342
ADR: 3,II
Merck Index: 13,05152

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point -118 °C
Molar Mass 137.01 g/mol
Boiling Point 92 °C (1013 hPa)
Vapor Pressure 41 hPa (20 °C)
Flash Point 19 °C
Refractive Index 1.435 (20 °C, 589 nm)
Density 1.26 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR