tert-Butylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01546.0100 8.01546.0500 8.01546.1000 8.01546.0100 8.01546.0500 8.01546.1000
EM8.01546.0100CS 970.12 CAD
EM8.01546.0100 EM8.01546.0500 EM8.01546.1000 CA8.01546.0100 CA8.01546.0500 CA8.01546.1000
tert-Butylamine for synthesis, Sigma-Aldrich®
tert-Butylamine
Formula: C₄H₁₁N
Boiling Pt: 44.4 °C (1013 hPa)
Melting Pt: –72 °C
Density: 0.6965 g/cm³ (20 °C)
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8 (3),I
Merck Index: 13,01544

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -72 °C
Molar Mass 73.14 g/mol
Boiling Point 44 °C
Vapor Pressure 300 hPa (20 °C)
Flash Point -38 °C DIN 51755 Part 1
Refractive Index 1.37 (20 °C, 589 nm)
Explosion Limit 1.5 - 9.2 %(V)
Density 0.69 g/cm3 (20 °C)
pH 12 (100 g/l, H2O, 20 °C)
Ignition Point 375 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR