Silver(I) acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Silver acetate, Acetic acid silver (I) salt

M-Clarity™ quality level = MQ200

8.01504.0005 8.01504.0025 8.01504.0100 8.01504.0005 8.01504.0025 8.01504.0100
EM8.01504.0005CS 200939.66 CAD
EM8.01504.0005 EM8.01504.0025 EM8.01504.0100 CA8.01504.0005 CA8.01504.0025 CA8.01504.0100
Silver(I) acetate for synthesis, Sigma-Aldrich®
Silver (I) acetate
Formula: C₂H₃AgO₂
Melting Pt: 2000 °C
Density: 3.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012446
CAS Number: 563-63-3
EINECS: 209-254-9
UN: 1759
ADR: 8,II
Merck Index: 12,08648

Order Now

Specification Test Results

Solubility 10.2 g/l (20 °C)
Molar Mass 166.91 g/mol
Density 3.26 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR