2-Ethylbutyric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01480.0500 8.01480.0100 8.01480.0500
EM8.01480.0100CS 1442.21 CAD
EM8.01480.0100 EM8.01480.0500 CA8.01480.0100 CA8.01480.0500
2-Ethylbutyric acid for synthesis, Sigma-Aldrich®
2-Ethylbutyric acid
Formula: C₆H₁₂O₂
Boiling Pt: 193…194 °C (1013 hPa)
Melting Pt: –14 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002670
CAS Number: 88-09-5
EINECS: 201-796-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03137

Order Now

Specification Test Results

Solubility 18 g/l (20 °C)
Melting Point -14 °C
Molar Mass 116.15 g/mol
Boiling Point 192 - 194 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 87 °C
Explosion Limit 1.4 %(V)
Density 0.92 g/cm3 (20 °C)
pH 3 (18 g/l, H2O, 20 °C)
Ignition Point 345 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR