AMP (2-Amino-2-methylpropanol) 93,0-98,0 % (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP

M-Clarity™ quality level = MQ200

8.01465.0250 8.01465.1000 8.01465.2500 8.01465.9026 8.01465.0250 8.01465.1000 8.01465.2500 8.01465.9026
EM8.01465.0250CS 1045.96 CAD
EM8.01465.0250 EM8.01465.1000 EM8.01465.2500 EM8.01465.9026 CA8.01465.0250 CA8.01465.1000 CA8.01465.2500 CA8.01465.9026
AMP (2-Amino-2-methylpropanol) 93,0-98,0 % (by GC) for synthesis, Sigma-Aldrich®
AMP (2-Amino-2-methylpropanol)
Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165 °C (1013 hPa (anhydrous))
Melting Pt: 30…31 °C
Density: 0.95 g/cm3 (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448

Order Now

Specification Test Results

Melting Point 30 - 31 °C
Molar Mass 89.14 g/mol
Boiling Point 165 °C (1013 hPa) (anhydrous)
Vapor Pressure 0.45 hPa (20 °C)
Flash Point 77.8 °C
Refractive Index 1.448 (20 °C, 589 nm)
Density 0.95 g/cm3 (25 °C)
pH 11.3 (10 g/l, H2O)
Ignition Point 438 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR