2-Amino-2-methyl-1,3-propanediol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.01464.0050 8.01464.0250 8.01464.0050 8.01464.0250
EM8.01464.0050CS 18038.86 CAD
EM8.01464.0050 EM8.01464.0250 CA8.01464.0050 CA8.01464.0250
2-Amino-2-methyl-1,3-propanediol for synthesis, Sigma-Aldrich®
2-Amino-2-methyl-1,3-propanediol
Formula: C₄H₁₁NO₂
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 102…105 °C
Density: 1.211 g/cm³ (20 °C)
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
UN: 3259
ADR: 8,III
Merck Index: 13,00447

Order Now

Specification Test Results

Solubility 2500 g/l (20 °C)
Melting Point 102 - 105 °C
Molar Mass 105.13 g/mol
Boiling Point 150 - 152 °C (13 hPa)
Vapor Pressure 13.3 hPa (152 °C)
Flash Point 100 °C
pH 10 - 11 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR