Bis(2-ethylhexyl)adipate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.01453.0100 8.01453.1000 8.01453.9025 8.01453.0100 8.01453.1000 8.01453.9025
EM8.01453.0100CS 2960.92 CAD
EM8.01453.0100 EM8.01453.1000 EM8.01453.9025 CA8.01453.0100 CA8.01453.1000 CA8.01453.9025
Bis(2-ethylhexyl)adipate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Bis(2-ethylhexyl) adipate
Formula: C₂₂H₄₂O₄
Boiling Pt: 247…252 °C (27 hPa)
Melting Pt: –68…–66 °C
Density: 0.93 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (20 °C)
Melting Point -68 - -66 °C
Molar Mass 370.57 g/mol
Boiling Point 247 - 252 °C (27 hPa)
Vapor Pressure  - 0.001 hPa (20 °C)
Flash Point 195 °C
Refractive Index 1.447 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Ignition Point 350 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR