2-Ethylaminoethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01392.0250 8.01392.1000 8.01392.0250 8.01392.1000
EM8.01392.0250CS 1148.66 CAD
EM8.01392.0250 EM8.01392.1000 CA8.01392.0250 CA8.01392.1000
2-Ethylaminoethanol for synthesis, Sigma-Aldrich®
2-Ethylaminoethanol
Formula: C₄H₁₁NO
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.914 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002841
CAS Number: 110-73-6
EINECS: 203-797-5
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -9 °C
Molar Mass 89.14 g/mol
Boiling Point 167 °C
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 78 °C
Refractive Index 1.4402 (20 °C, 589 nm)
Explosion Limit 1.6 - 11.7 %(V)
Density 0.914 g/cm3 (20 °C)
pH 12.3 (100 g/l, H2O, 20 °C)
Ignition Point 330 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR