Acetal for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,1-Diethoxyethane, Acetaldehyde diethyl acetal

M-Clarity™ quality level = MQ200

8.01366.0005 8.01366.0250 8.01366.0005 8.01366.0250
EM8.01366.0005CS 3929.46 CAD
EM8.01366.0005 EM8.01366.0250 CA8.01366.0005 CA8.01366.0250
Acetal for synthesis, Sigma-Aldrich®
Acetal
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 102.7 °C (1013 hPa)
Melting Pt: –100 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: –21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009243
CAS Number: 105-57-7
EINECS: 203-310-6
UN: 1088
ADR: 3,II
Merck Index: 13,00040

Order Now

Specification Test Results

Solubility 46 g/l (25 °C)
Melting Point -100 °C
Molar Mass 118.17 g/mol
Boiling Point 102 °C (1013 hPa)
Vapor Pressure 20.3 hPa (20 °C)
Flash Point -21 °C
Refractive Index 1.3825 (20 °C, 589 nm)
Explosion Limit 1.6 - 10.4 %(V)
Density 0.83 g/cm3 (20 °C)
Ignition Point 230 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR