2-Aminopyridine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.01113.0100
8.01113.0500
8.01113.0100
8.01113.0500
EM8.01113.0100CS
832.66
CAD
EM8.01113.0100
EM8.01113.0500
CA8.01113.0100
CA8.01113.0500
2-Aminopyridine for synthesis, Sigma-Aldrich®
2-Aminopyridine
Formula:
C₅H₆N₂ Boiling Pt: 210.6 °C (1013 hPa) Melting Pt: 50…52 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006312 CAS Number: 504-29-0 EINECS: 207-988-4 UN: 2671 ADR: 6.1,II Merck Index: 13,00472 |
Specification Test Results
Solubility | 890 g/l (21 °C) |
Melting Point | 55 - 58 °C |
Molar Mass | 94.11 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 209 - 211 °C (1013 hPa) |
Vapor Pressure | 5 hPa (125 °C) |
Flash Point | 92 °C |
pH | (H2O, 20 °C) alkaline,Aqueous solution |
Ignition Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...