9-Aminoacridine hydrochloride monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 9-Aminoacridine hydrochloride hydrate, Acridin-9-amine hydrochloride hydrate

M-Clarity™ quality level = MQ200

8.01112.0025 8.01112.0100 8.01112.0025 8.01112.0100
EM8.01112.0025CS 18512.86 CAD
EM8.01112.0025 EM8.01112.0100 CA8.01112.0025 CA8.01112.0100
9-Aminoacridine hydrochloride monohydrate for synthesis, Sigma-Aldrich®
9-Aminoacridine hydrochloride monohydrate
Formula: C₁₃H₁₁ClN₂·1H₂O
MDL Number: MFCD00150071
CAS Number: 52417-22-8
EINECS: 205-145-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 3.3 g/l
Melting Point 240 - 244 °C
Molar Mass 248.71 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR