Trimethyl orthoformate zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


MQ200

8.00885.0250 8.00885.1000 8.00885.0250 8.00885.1000
EM8.00885.0250CS 791.58 CAD
EM8.00885.0250 EM8.00885.1000 CA8.00885.0250 CA8.00885.1000
Trimethyl orthoformate zS, Sigma-Aldrich®
Trimethyl orthoformate
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 101
Melting Pt: -53
Density: 0.9691 g/cm³ (20 °C)
Flash Pt: 13
MDL Number: MFCD00008483
CAS Number: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3 II
Merck Index: 12,07012

Order Now

Specification Test Results

Solubility 10 g/l (20 °C) Hydrolysis
Melting Point -53 °C
Molar Mass 106.12 g/mol
Boiling Point 102 °C (1013 hPa)
Vapor Pressure 10 hPa (7 °C)
Flash Point 13 °C
Refractive Index 1.379 (20 °C, 589 nm)
Explosion Limit 1.4 - 44.6 %(V)
Density 0.97 g/cm3 (20 °C)
Ignition Point 255 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR