Ethylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Aminoethane hydrochloride, Ethylamine hydrochloride

M-Clarity™ quality level = MQ200

8.00874.0250 8.00874.1000 8.00874.0250 8.00874.1000
EM8.00874.0250CS 1358.8 CAD
EM8.00874.0250 EM8.00874.1000 CA8.00874.0250 CA8.00874.1000
Ethylammonium chloride for synthesis, Sigma-Aldrich®
Ethylammonium chloride
Formula: C₂H₈ClN
MDL Number: MFCD00012885
CAS Number: 557-66-4
EINECS: 209-182-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 107 - 108 °C
Molar Mass 81.54 g/mol

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR