Valeric acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Pentanoic acid
M-Clarity™ quality level = MQ200
8.00821.0100
8.00821.0500
8.00821.1000
8.00821.0100
8.00821.0500
8.00821.1000
EM8.00821.0100CS
900.6
CAD
EM8.00821.0100
EM8.00821.0500
EM8.00821.1000
CA8.00821.0100
CA8.00821.0500
CA8.00821.1000
Valeric acid for synthesis, Sigma-Aldrich®
Valeric acid
Formula:
C₅H₁₀O₂ Boiling Pt: 186 °C (1013 hPa) Melting Pt: –32 °C Density: 1.11 g/cm³ (20 °C) Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004413 CAS Number: 109-52-4 EINECS: 203-677-2 UN: 3265 ADR: 8,I Merck Index: 12,10042 |
Specification Test Results
Solubility | 40 g/l (20 °C) |
Melting Point | -32 °C |
Molar Mass | 102.13 g/mol |
Boiling Point | 186 °C (1013 hPa) |
Vapor Pressure | 0.3 hPa (20 °C) |
Flash Point | 86 °C |
Refractive Index | 1.4076 (20 °C, 589 nm) |
Explosion Limit | 1.8 - 7.3 %(V) |
Density | 0.938 g/cm3 (20 °C) |
pH | 2.7 (40 g/l, H2O, 20 °C) |
Ignition Point | 360 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...