Undec-10-enoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00813.0500 8.00813.5000 8.00813.0500 8.00813.5000
EM8.00813.0500CS 3727.22 CAD
EM8.00813.0500 EM8.00813.5000 CA8.00813.0500 CA8.00813.5000
Undec-10-enoic acid for synthesis, Sigma-Aldrich®
Undec-10-enoic acid
Formula: C₁₁H₂₀O₂
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 24 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 146 °C
MDL Number: MFCD00004442
CAS Number: 112-38-9
EINECS: 203-965-8
Merck Index: 12,09983

Order Now

Specification Test Results

Solubility 73.7 mg/l (30 °C)
Melting Point 24 °C
Molar Mass 184.27 g/mol
Boiling Point 275 °C (1013 mbar)
Flash Point 149 °C
Density 0.91 g/cm3 (20 °C)
Ignition Point 275 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR