Pyromellitic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,2,4,5-Benzenetetracarboxylic acid, Benzene-1,2,4,5-tetracarboxylic acid

M-Clarity™ quality level = MQ200

8.00784.0005 8.00784.0050 8.00784.0250 8.00784.0005 8.00784.0050 8.00784.0250
EM8.00784.0005CS 58017.6 CAD
EM8.00784.0005 EM8.00784.0050 EM8.00784.0250 CA8.00784.0005 CA8.00784.0050 CA8.00784.0250
Pyromellitic acid for synthesis, Sigma-Aldrich®
Pyromellitic acid
Formula: C₁₀H₆O₈
MW: 254.15 g/mol
Boiling Pt: 516
Melting Pt: 272
Density: 1.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002471
CAS Number: 89-05-4
EINECS: 201-879-5
Merck Index: 12,08188

Order Now

Specification Test Results

Solubility 15 g/l (25 °C)
Melting Point 250 - 260 °C
Molar Mass 254.15 g/mol
Density 1.79 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR