2-Ethylcaproic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Ethylhexanoic acid

M-Clarity™ quality level = MQ200

8.00758.0100 8.00758.1000 8.00758.0100 8.00758.1000
EM8.00758.0100CS 970.12 CAD
EM8.00758.0100 EM8.00758.1000 CA8.00758.0100 CA8.00758.1000
2-Ethylcaproic acid for synthesis, Sigma-Aldrich®
2-Ethylcaproic acid
Formula: C₈H₁₆O₂
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: –82.8 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002675
CAS Number: 149-57-5
EINECS: 205-743-6

Order Now

Specification Test Results

Solubility 1.4 g/l (20 °C)
Melting Point -59 °C
Molar Mass 144.21 g/mol
Boiling Point 226 - 229 °C (1013 hPa)
Vapor Pressure 0.04 hPa (20 °C)
Flash Point 114 °C
Refractive Index 1.4241 (20 °C, 589 nm)
Explosion Limit 0.9 - 6.7 %(V)
Density 0.91 g/cm3 (20 °C)
pH 3 (1.4 g/l, H2O, 20 °C)
Ignition Point 310 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR