1,2,3,6-Tetrahydrophthalic anhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00742.0100 8.00742.1000 8.00742.9025 8.00742.0100 8.00742.1000 8.00742.9025
EM8.00742.0100CS 7491.6 CAD
EM8.00742.0100 EM8.00742.1000 EM8.00742.9025 CA8.00742.0100 CA8.00742.1000 CA8.00742.9025
1,2,3,6-Tetrahydrophthalic anhydride for synthesis, Sigma-Aldrich®
1,2,3,6-Tetrahydrophthalic anhydride
Formula: C₈H₈O₃
Melting Pt: 101.4 °C
Density: 1.36 g/cm3 (20 °C)
Storage Temperature: Ambient
CAS Number: 85-43-8
EINECS: 201-605-4
UN: 2698
ADR: 8,III

Order Now

Specification Test Results

Solubility 10 g/l (20 °C)
Melting Point 103 °C
Molar Mass 152.14 g/mol
Boiling Point 195 °C (67 hPa)
Vapor Pressure 0.013 hPa (20 °C)
Flash Point 156 °C
Density 1.375 g/cm3 (20 °C)
pH 2.1 (10 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR