Succinyl chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Succinyl dichloride
M-Clarity™ quality level = MQ200
8.00678.0050
8.00678.0050
EM8.00678.0050CS
19735.48
CAD
EM8.00678.0050
CA8.00678.0050
Succinyl chloride for synthesis, Sigma-Aldrich®
Succinyl chloride
Formula:
C₄H₄Cl₂O₂ |
MDL Number:
MFCD00000749 CAS Number: 543-20-4 UN: 3265 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 20 °C |
Molar Mass | 154.98 g/mol |
Boiling Point | 190 °C |
Flash Point | 76 °C |
Refractive Index | 1.4691 (20 °C, 589 nm) |
Density | 1.38 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...