2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
8.00610.0250
8.00610.1000
8.00610.0250
8.00610.1000
EM8.00610.0250CS
559.32
CAD
EM8.00610.0250
EM8.00610.1000
CA8.00610.0250
CA8.00610.1000
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
2-Hydroxypropyl methacrylate (mixture of isomers, methacrylic acid, monoester with propane-1,2-diol)
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004536 CAS Number: 27813-02-1 EINECS: 248-666-3 |
Specification Test Results
Assay isomers (GC, area%) | ≥ 97.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 1.027 - 1.030 |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...