Methacrylic acid alkylester (mixture of C₁₂-C₁₆) stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00589.0100 8.00589.0500 8.00589.0100 8.00589.0500
EM8.00589.0100CS 936.94 CAD
EM8.00589.0100 EM8.00589.0500 CA8.00589.0100 CA8.00589.0500
Methacrylic acid alkylester (mixture of C₁₂-C₁₆) stabilized for synthesis, Sigma-Aldrich®
Methacrylic acid alkylester (mixture of C₁₂-C₁₆)
Storage Temperature: Ambient
CAS Number: 90551-76-1
EINECS: 292-094-7

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -25 °C
Boiling Point 300 °C
Flash Point 105 °C
Density 0.87 g/cm3 (20 °C)
assay n-C12 isomer (GC, area%) 60.0 - 70.0 % (a/a)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR