Ethyl methacrylate stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester, Ethyl-2-methylprop-2-enoat, Ethyl-2-methylpropenoat, Ethyl 2-methylprop-2-enoate

M-Clarity™ quality level = MQ200

8.00579.0100 8.00579.0500 8.00579.0100 8.00579.0500
EM8.00579.0100CS 829.5 CAD
EM8.00579.0100 EM8.00579.0500 CA8.00579.0100 CA8.00579.0500
Ethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Ethyl methacrylate
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 100
Melting Pt: -48
Density: 0.9158 g/cm³ (20 °C)
Flash Pt: 20
MDL Number: MFCD00009161
CAS Number: 97-63-2
EINECS: 202-597-5
UN: 2277
ADR: 3 II

Order Now

Specification Test Results

Solubility 5.1 g/l (20 °C)
Melting Point -61 - -59 °C
Molar Mass 114.14 g/mol
Boiling Point 119 - 121 °C (1013 hPa)
Vapor Pressure 21.3 hPa (20 °C)
Flash Point 18 °C
Refractive Index 1.4128 (20 °C, 589 nm)
Explosion Limit 1.8 %(V)
Density 0.913 g/cm3 (20 °C)
Ignition Point 329 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR