Triphenyl phosphite for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00551.0100 8.00551.1000 8.00551.0100 8.00551.1000
EM8.00551.0100CS 1200.8 CAD
EM8.00551.0100 EM8.00551.1000 CA8.00551.0100 CA8.00551.1000
Triphenyl phosphite for synthesis, Sigma-Aldrich®
Triphenyl phosphite
Formula: C₁₈H₁₅O₃P
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 22…25 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) (slow decomposition)
Melting Point 22 - 25 °C
Molar Mass 310.29 g/mol
Boiling Point 360 °C (1013 hPa) decomposes
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 188 °C
Refractive Index 1.5903 (20 °C, 589 nm)
Density 1.19 g/cm3 (20 °C)
pH 1 (200 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR