Butyric anhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00503.0100 8.00503.1000 8.00503.0100 8.00503.1000
EM8.00503.0100CS 973.28 CAD
EM8.00503.0100 EM8.00503.1000 CA8.00503.0100 CA8.00503.1000
Butyric anhydride for synthesis, Sigma-Aldrich®
Butyric anhydride
Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 198
Melting Pt: -73
Density: 0.963 g/cm³ (20 °C)
Flash Pt: 83
Storage Temperature: Ambient
MDL Number: MFCD00009389
CAS Number: 106-31-0
EINECS: 203-383-4
UN: 2739
ADR: 8 III
Merck Index: 13,01594

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point -75 °C
Molar Mass 158.19 g/mol
Boiling Point 195 - 199 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.413 (20 °C, 589 nm)
Explosion Limit 1.1 - 7.6 %(V)
Density 0.963 g/cm3 (20 °C)
Ignition Point 290 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR