Isobutyric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00472.0100 8.00472.1000 8.00472.0100 8.00472.1000
EM8.00472.0100CS 1216.6 CAD
EM8.00472.0100 EM8.00472.1000 CA8.00472.0100 CA8.00472.1000
Isobutyric acid for synthesis, Sigma-Aldrich®
Isobutyric acid
Formula: C₄H₈O₂
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002658
CAS Number: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3 (8),III
Merck Index: 13,05172

Order Now

Specification Test Results

Solubility 210 g/l (20 °C)
Melting Point -45 °C
Molar Mass 88.11 g/mol
Boiling Point 153 - 155 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 57.5 °C
Refractive Index 1.3925 (20 °C, 589 nm)
Explosion Limit 2 - 12 %(V)
Density 0.95 g/cm3 (20 °C)
Ignition Point 420 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR