trans-Anethole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00429.0100 8.00429.0250 8.00429.0100 8.00429.0250
EM8.00429.0100CS 3441.24 CAD
EM8.00429.0100 EM8.00429.0250 CA8.00429.0100 CA8.00429.0250
trans-Anethole for synthesis, Sigma-Aldrich®
trans-Anethole
Formula: C₁₀H₁₂O
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 21…23 °C
Density: 0.9883 g/cm³ (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00009284
CAS Number: 4180-23-8
EINECS: 224-052-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 21 - 23 °C
Molar Mass 148.20 g/mol
Boiling Point 235 - 237 °C (1013 hPa)
Vapor Pressure 1.33 hPa (63 °C)
Flash Point 96 °C
Refractive Index 1.56 (20 °C, 589 nm)
Density 0.99 g/cm3 (20 °C)
pH 7 (H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR