3-Aminopropan-1-ol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00423.0005 8.00423.0250 8.00423.9025 8.00423.0005 8.00423.0250 8.00423.9025
EM8.00423.0005CS 2423.72 CAD
EM8.00423.0005 EM8.00423.0250 EM8.00423.9025 CA8.00423.0005 CA8.00423.0250 CA8.00423.9025
3-Aminopropan-1-ol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
3-Aminopropan-1-ol

Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Formula: C₃H₉NO
Boiling Pt: 187…190 °C
Melting Pt: 11.5…12.4 °C
Density: 0.99 g/cm3 (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00008223
CAS Number: 156-87-6
EINECS: 205-864-4
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Assay (GC, area%) ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.986 - 0.988
Water (K. F.) ≤ 0.30 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR