3-Chloropropene Stabilised zS, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
3-Chloro-1-propene, Allyl chloride
Stabilization:
Propylenoxid
MQ200
8.00257.0100
8.00257.1000
8.00257.0100
8.00257.1000
EM8.00257.0100CS
1124.96
CAD
EM8.00257.0100
EM8.00257.1000
CA8.00257.0100
CA8.00257.1000
3-Chloropropene Stabilised zS, Sigma-Aldrich®
3-Chloropropene
Formula:
C₃H₅Cl MW: 76.53 g/mol Boiling Pt: 44 Melting Pt: -136 Density: 0.93 g/cm³ (20 °C) Flash Pt: –32 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000984 CAS Number: 107-05-1 EINECS: 203-457-6 UN: 1100 ADR: 3 (6.1) I Merck Index: 13,00286 |
Specification Test Results
Solubility | 3.6 g/l (20 °C) |
Melting Point | -136 °C |
Molar Mass | 76.52 g/mol |
Boiling Point | 45 °C (1013 hPa) |
Vapor Pressure | 400 hPa (20 °C) |
Explosion Limit | 3.3 - 11.2 %(V) |
Density | 0.94 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 390 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...