2-Acetylthiophene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Thiophen-2-yl-ethanone, 1-(2-Thienyl)-1-ethanone, Methyl 2-thienyl ketone

M-Clarity™ quality level = MQ200

8.00175.0050 8.00175.0250 8.00175.0050 8.00175.0250
EM8.00175.0050CS 13218.28 CAD
EM8.00175.0050 EM8.00175.0250 CA8.00175.0050 CA8.00175.0250
2-Acetylthiophene for synthesis, Sigma-Aldrich®
2-Acetylthiophene
Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.1704 g/cm³ (20 °C)
MDL Number: MFCD00005442
CAS Number: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 14 g/l (30 °C)
Melting Point 9 - 11 °C
Molar Mass 126.18 g/mol
Boiling Point 211 - 213 °C (1013 hPa)
Flash Point 97 °C
Density 1.17 g/cm3 (20 °C)
Ignition Point 265 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR