6-Aminocaproic acid ≥99% (by perchloric acid titration) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

M-Clarity™ quality level = MQ200

8.00145.0005 8.00145.0250 8.00145.1000 8.00145.5000 8.00145.9010 8.00145.0005 8.00145.0250 8.00145.1000 8.00145.5000 8.00145.9010
EM8.00145.0005CS 48912.06 CAD
EM8.00145.0005 EM8.00145.0250 EM8.00145.1000 EM8.00145.5000 EM8.00145.9010 CA8.00145.0005 CA8.00145.0250 CA8.00145.1000 CA8.00145.5000 CA8.00145.9010
6-Aminocaproic acid ≥99% (by perchloric acid titration) for synthesis, Sigma-Aldrich®
6-Aminocaproic acid
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 205 °C
Density: 1.1313 g/cm3 (20 °C)
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
EINECS: 200-469-3
Merck Index: 13,00431

Order Now

Specification Test Results

Solubility 800 g/l (20 °C)
Molar Mass 131.17 g/mol
Flash Point 207 - 209 °C
pH 7.0 - 7.5 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR