Acetonylacetone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00105.0100 8.00105.0250 8.00105.0100 8.00105.0250
EM8.00105.0100CS 2945.12 CAD
EM8.00105.0100 EM8.00105.0250 CA8.00105.0100 CA8.00105.0250
Acetonylacetone for synthesis, Sigma-Aldrich®
Acetonylacetone
Formula: C₆H₁₀O₂
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: –5.4 °C
Density: 0.971 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008792
CAS Number: 110-13-4
EINECS: 203-738-3
Merck Index: 13,00072

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -5.4 °C
Molar Mass 114.14 g/mol
Boiling Point 85 - 87 °C (26.6 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.423 (20 °C, 589 nm)
Explosion Limit 1.5 %(V)
Density 0.974 g/cm3 (20 °C)
pH 6.1 (10 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR