Acetylacetone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: ACAC, 2,4-Pentanedione

M-Clarity™ quality level = MQ200

8.00023.0005 8.00023.0250 8.00023.9010 8.00023.0005 8.00023.0250 8.00023.1000 8.00023.9010
EM8.00023.0005CS 2760.26 CAD
EM8.00023.0005 EM8.00023.0250 EM8.00023.9010 CA8.00023.0005 CA8.00023.0250 CA8.00023.1000 CA8.00023.9010
Acetylacetone for synthesis, Sigma-Aldrich®
Acetylacetone
Formula: CH₃COCH₂COCH₃
MW: 100.12 g/mol
Boiling Pt: 140.4
Melting Pt: -23
Density: 0.96945 g/cm³ (20 °C)
Flash Pt: 30
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3 (6.1) III
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c85a64bf9c7997e9ab03e421a93e7c71e57b8a3&vid=134ed3029693deef3759692be256c57b3a58b087&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 200 g/l (20 °C)
Melting Point -23 °C
Molar Mass 100.12 g/mol
Boiling Point 138 - 140 °C (1013 hPa)
Vapor Pressure 9 hPa (20 °C)
Flash Point 34 °C
Explosion Limit 2.4 - 11.4 %(V)
Density 0.97 g/cm3 (20 °C)
pH 6 (200 g/l, H2O, 20 °C)
Ignition Point 340 °C
Evaporation Number 20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR