Osmium tetraoxide for microscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid

1.24505.0001 1.24505.0500
EM1.24505.0001CS 88679.08 CAD
EM1.24505.0001 EM1.24505.0500 CA1.24505.0001 CA1.24505.0500
Osmium tetraoxide for microscopy, Sigma-Aldrich®
Osmium tetraoxide
Formula: O₄Os
MW: 254.23 g/mol
Boiling Pt: 130
Melting Pt: 40
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 2471
ADR: 6.1
Merck Index: 13,06962

Order Now

Specification Test Results

Solubility 65 g/l (20 °C)
Melting Point 40 °C
Molar Mass 254.19 g/mol
Bulk Density 1500 kg/m3
Boiling Point 130 °C (1013 hPa)
Vapor Pressure 10 hPa (20 °C)
Density 4.91 g/cm3 (20 °C)
pH 6 (25 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR