Palladium ≥99%, powder, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ300

1.19225.0001 1.19225.0005 1.19225.0001 1.19225.0005
EM1.19225.0001CS 42034.32 CAD
EM1.19225.0001 EM1.19225.0005 CA1.19225.0001 CA1.19225.0005
Palladium ≥99%, powder, EMSURE® for analysis, Supelco®
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
ADR: 4.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1554 °C
Molar Mass 106.4 g/mol
Boiling Point 2200 - 2900 °C
Density 12.02 g/cm3 (20 °C)
pH (H2O) not applicable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR