Silver(I) oxide ≥99%, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.19208.0025 1.19208.0100 1.19208.0025 1.19208.0100
EM1.19208.0025CS 120214.3 CAD
EM1.19208.0025 EM1.19208.0100 CA1.19208.0025 CA1.19208.0100
Silver(I) oxide ≥99%, Supelco®
Silver(I) oxide
Formula: Ag₂O
MDL Number: MFCD00003404
CAS Number: 20667-12-3
UN: 3085
ADR: 5.1 (8),

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 200 °C decomposes
Molar Mass 231.74 g/mol
Bulk Density 950 kg/m3
Density 7.2 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR