Silver chloride ≥99%, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ300
1.19203.0025
1.19203.0100
1.19203.0025
1.19203.0100
EM1.19203.0025CS
150438.12
CAD
EM1.19203.0025
EM1.19203.0100
CA1.19203.0025
CA1.19203.0100
Silver chloride ≥99%, Supelco®
Silver chloride
Formula:
AgCl MW: 143.32 g/mol |
MDL Number:
MFCD00003399 CAS Number: 7783-90-6 UN: 3077 ADR: 9, |
Specification Test Results
Solubility | 0.00188 g/l (25 °C) |
Melting Point | 455 °C |
Molar Mass | 143.32 g/mol |
Boiling Point | 1554 °C (1013 hPa) |
Density | 5.56 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...