Test Kits, Nitrate, Spectroquant®, Supelco®
Supplier: MilliporeSigma
1.14556.0001 1.14764.0001 1.14773.0001 1.09713.0001 1.14764.0001 1.14773.0001
EM1.09713.0001CS 4476.14 CAD
EM1.09713.0001 EM1.14556.0001 EM1.14764.0001 EM1.14773.0001 CA1.09713.0001 CA1.14556.0001 CA1.14764.0001 CA1.14773.0001 CA1.09713.0002
Test Kits, Nitrate, Spectroquant®, Supelco®
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
Spectroquant® cell test kits
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.