Ammonium Cell Tests, Method: photometric, Spectroquant®, Supelco®
Supplier: MilliporeSigma
1.14544.0001 1.14558.0001 1.14559.0001 1.14739.0001 1.14544.0001 1.14558.0001 1.14559.0001 1.14739.0001
EM1.14544.0001CS 1216.6 CAD
EM1.14544.0001 EM1.14558.0001 EM1.14559.0001 EM1.14739.0001 CA1.14544.0001 CA1.14558.0001 CA1.14559.0001 CA1.14739.0001
Ammonium Cell Tests, Method: photometric, Spectroquant®, Supelco®
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).