Chlorine Test (Free Chlorine) Method: Colorimetric, DPD, with Color Card and Comparator Block with Long Test Tubes 0.010 - 0.025 - 0.045 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.30 mg/L Cl₂ MColortest™
Supplier: MilliporeSigma
1.14434.0001 1.14434.0001
EM1.14434.0001CS 5956.6 CAD
EM1.14434.0001 CA1.14434.0001
Chlorine Test (Free Chlorine) Method: Colorimetric, DPD, with Color Card and Comparator Block with Long Test Tubes 0.010 - 0.025 - 0.045 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.30 mg/L Cl₂ MColortest™
Environmental Test Kits