Tin (II) sulfate, LAB., Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.12416.0250 1.12416.1000 1.12416.0250 1.12416.1000
EM1.12416.0250CS 3588.18 CAD
EM1.12416.0250 EM1.12416.1000 CA1.12416.0250 CA1.12416.1000
Tin (II) sulfate, LAB., Sigma-Aldrich®
Tin (II) sulphate
Formula: O₄SSn
MDL Number: MFCD00011246
CAS Number: 7488-55-3

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 214.77 g/mol
Bulk Density 800 kg/m3
pH 1.6 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR