Titanium ≥98%, fine powder, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.12379.0250
1.12379.0250
EM1.12379.0250CS
2839.81
CAD
EM1.12379.0250
CA1.12379.0250
Titanium ≥98%, fine powder, Sigma-Aldrich®
Titanium
Formula:
Ti MW: 47.88 g/mol |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 UN: 2546 ADR: 4.2, |
Specification Test Results
Solubility | (20 °C) insoluble,(reaction) |
Melting Point | 1670 °C |
Molar Mass | 47.88 g/mol |
Bulk Density | 1260 kg/m3 |
Boiling Point | 3260 °C |
Density | 4.5 g/cm3 (20 °C) |
pH | 5 - 8 (50 g/l, H2O) suspension |
Ignition Point | 300 - 450 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...