Nickel 99, powder, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
MQ200
1.12277.0250
1.12277.1000
1.12277.0250
1.12277.1000
EM1.12277.0250CS
20931.84
CAD
EM1.12277.0250
EM1.12277.1000
CA1.12277.0250
CA1.12277.1000
Nickel 99, powder, Sigma-Aldrich®
Nickel
Formula:
Ni MW: 58.69 g/mol Boiling Pt: 3007 Melting Pt: 1453 |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 UN: 3089 ADR: 4.1 III Merck Index: 12,06582 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 1453 °C |
Molar Mass | 58.69 g/mol |
Bulk Density | 1600 - 2600 kg/m3 |
Boiling Point | 2832 °C |
Density | 8.90 g/cm3 (25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...