Lithium nitrate 98, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
MQ200
1.12230.0250
1.12230.1000
1.12230.0250
1.12230.1000
EM1.12230.0250CS
8961.76
CAD
EM1.12230.0250
EM1.12230.1000
CA1.12230.0250
CA1.12230.1000
Lithium nitrate 98, Sigma-Aldrich®
Lithium nitrate
Formula:
LiNO₃ MW: 68.95 g/mol Melting Pt: 255 |
MDL Number:
MFCD00011094 CAS Number: 7790-69-4 UN: 2722 ADR: 5.1 Merck Index: 13,05558 |
Specification Test Results
Solubility | 522 g/l (20 °C) |
Melting Point | 255 °C |
Molar Mass | 68.94 g/mol |
Bulk Density | 910 kg/m3 |
Density | 2.36 g/cm3 (20 °C) |
pH | 7 - 9 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...