Lanthanum(III) oxide, Supelco®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
1.12220.0100
1.12220.0500
1.12220.0100
1.12220.0500
EM1.12220.0100CS
36951.46
CAD
EM1.12220.0100
EM1.12220.0500
CA1.12220.0100
CA1.12220.0500
Lanthanum(III) oxide, Supelco®
Lanthanum (III) oxide
EMD # 1.12220.0500 replaces and is identical to EMD # LX0045-2.
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 |
Specification Test Results
Melting Point | 2315 °C |
Molar Mass | 325.82 g/mol |
Bulk Density | 1300 kg/m3 |
Boiling Point | 4200 °C |
Density | 6.51 g/cm3 (20 °C) |
pH | 9.0 (50 g/l, H2O, 20 °C) (slurry) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...