Boron, powder, Sigma-Aldrich®

Supplier: MilliporeSigma

1.12070.0025 1.12070.1000 1.12070.0025 1.12070.1000
EM1.12070.0025CS 7274.32 CAD
EM1.12070.0025 EM1.12070.1000 CA1.12070.0025 CA1.12070.1000
Boron, powder, Sigma-Aldrich®
Boron
Formula: B
MW: 10.81 g/mol
MDL Number: MFCD00134034
CAS Number: 7440-42-8
UN: 3178
Merck Index: 13,01333

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 2075 °C
Molar Mass 10.81 g/mol
Bulk Density 200 - 300 kg/m3
Boiling Point 4000 °C
Density 1.73 g/cm3 (15 °C)
Ignition Point 700 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR